5-(Trifluoromethoxy)salicylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzoic acid
PC5255-1G PC5255-10G PC5255-5G
APOSPC5255-1GEA 42.2 EUR
APOSPC5255-1G APOSPC5255-10G APOSPC5255-5G
5-(Trifluoromethoxy)salicylic acid
5-(Trifluoromethoxy)salicylic acid
Formula: C₈H₅F₃O₄
MW: 222.12 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD01091003
CAS Number: 129644-57-1