Undecafluoropentyl Iodide 98%
Supplier: Apollo Scientific
Warning
Synonyms: Perfluoropentyl Iodide
PC5264-25G PC5264-5G
APOSPC5264-25GEA 284 EUR
APOSPC5264-25G APOSPC5264-5G
Undecafluoropentyl Iodide 98%
Undecafluoropentyl Iodide
Formula: C₅F₁₁I
MW: 395.94 g/mol
Boiling Pt: 94.4 °C
Melting Pt: –50 °C
Density: 2.06 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08461629
CAS Number: 638-79-9