2-(Trifluoromethyl)pyridine
Supplier: Apollo Scientific
Danger
PC5273-1G PC5273-25G PC5273-5G
APOSPC5273-1GEA 21.9 EUR
APOSPC5273-1G APOSPC5273-25G APOSPC5273-5G
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 139…141 °C/754mmHg
Density: 1.275 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02183562
CAS Number: 368-48-9
UN: 1993
ADR: 3 III