2-Chloro-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC5289-500MG PC5289-5G
APOSPC5289-500MGEA 21.9 EUR
APOSPC5289-500MG APOSPC5289-5G
2-Chloro-3-(trifluoromethyl)benzaldehyde 98%
2-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
Melting Pt: 37…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00792414
CAS Number: 93118-03-7