N-Acetyl-5-bromo-2-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5'-Bromo-2'-fluoroacetanilide
PC53006-1G PC53006-5G
APOSPC53006-1GEA 158 EUR
APOSPC53006-1G APOSPC53006-5G
N-Acetyl-5-bromo-2-fluoroaniline 98%
N-Acetyl 5-bromo-2-fluoroaniline
Formula: C₈H₇BrFNO
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD12546480
CAS Number: 88288-12-4