3,4-Difluorothiophenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Difluorobenzene-1-thiol, 3,4-Difluorobenzenethiol, 1,2-Difluoro-4-mercaptobenzene, 3,4-Difluorothiophenol
PC5301-25G PC5301-5G
APOSPC5301-25GEA 430 EUR
APOSPC5301-25G APOSPC5301-5G
3,4-Difluorothiophenol 97%
3,4-Difluorothiophenol
Formula: C₆H₄F₂S
MW: 146.16 g/mol
Boiling Pt: 169…170 °C
Density: 1.323 g/cm³ (25 °C)
Flash Pt: 48.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094279
CAS Number: 60811-24-7
UN: 3336
ADR: 3,III