1-Bromo-4-difluoromethoxy-2-fluorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 1-bromo-4-(difluoromethoxy)-2-fluorobenzene
PC53076-1G PC53076-5G PC53076-10G
APOSPC53076-1GEA 261 EUR
APOSPC53076-1G APOSPC53076-5G APOSPC53076-10G
1-Bromo-4-difluoromethoxy-2-fluorobenzene
1-Bromo-4-difluoromethoxy-2-fluorobenzene
Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390897
CAS Number: 1242258-38-3