5-Bromo-3-fluoro-2-nitrotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-1-fluoro-3-methyl-2-nitrobenzene
PC53128-1G PC53128-250MG
APOSPC53128-1GEA 256 EUR
APOSPC53128-1G APOSPC53128-250MG
5-Bromo-3-fluoro-2-nitrotoluene
5-Bromo-3-fluoro-2-nitrotoluene
Formula: C₇H₅BrFNO₂
MW: 234.0225 g/mol
Melting Pt: 55…60 °C
Storage Temperature: Ambient
MDL Number: MFCD22566379
CAS Number: 1224629-03-1