2,6-Difluoro-4-iodophenol
Supplier: Apollo Scientific
Warning
PC53359-10G PC53359-1G
APOSPC53359-10GEA 379 EUR
APOSPC53359-10G APOSPC53359-1G
2,6-Difluoro-4-iodophenol
2,6-Difluoro-4-iodophenol
Formula: C₆H₃F₂IO
MW: 255.99 g/mol
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD09907951
CAS Number: 950858-06-7