4-(Trifluoromethyl)phenylhydrazinium chloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride
PC5337-25G PC5337-100G
APOSPC5337-25GEA 26.1 EUR
APOSPC5337-25G APOSPC5337-100G
4-(Trifluoromethyl)phenylhydrazinium chloride 97%
4-(Trifluoromethyl)phenylhydrazinium chloride
Formula: C₇H₈ClF₃N₂
MW: 212.6
Storage Temperature: Ambient
MDL Number: MFCD00204233
CAS Number: 2923-56-0