1H,1H,5H-Octafluoropentyl acrylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,3,3,4,4,5,5-Octafluoropentyl acrylate
PC5352-5G PC5352-1G PC5352-25G
APOSPC5352-5GEA 21.9 EUR
APOSPC5352-5G APOSPC5352-1G APOSPC5352-25G
1H,1H,5H-Octafluoropentyl acrylate 97%
1H,1H,5H-Octafluoropentyl acrylate
Formula: C₈H₆F₈O₂
MW: 286.12 g/mol
Boiling Pt: 56 °C/8mmHg
Density: 1.481 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039279
CAS Number: 376-84-1
UN: 3077
ADR: 9+EHS III