1H,1H,5H-Octafluoro-1-iodopentane 97%
Supplier: Apollo Scientific
Synonyms: 1H,1H,5H-Octafluoropentyl iodide
PC5352E-25G PC5352E-5G PC5352E-1G
APOSPC5352E-25GEA 607 EUR
APOSPC5352E-25G APOSPC5352E-5G APOSPC5352E-1G
1H,1H,5H-Octafluoro-1-iodopentane 97%
1H,1H,5H-Octafluoro-1-iodopentane
Formula: C₅H₃F₈I
MW: 341.97 g/mol
Boiling Pt: 136…137 °C
Density: 2.037 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039406
CAS Number: 678-74-0