3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
|
Warning
|
PC5361-1G
PC5361-250MG
PC5361-5G
APOSPC5361-1GEA
128
EUR
APOSPC5361-1G
APOSPC5361-250MG
APOSPC5361-5G
3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
3-Bromo-2-chloro-6-(trifluoromethyl)pyridine
Formula:
C₆H₂BrClF₃N Melting Pt: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD11226615 CAS Number: 1159512-34-1 |