1H,1H,7H-Dodecafluoroheptyl-p-toluenesulphonate 95%
Supplier: Apollo Scientific
Warning
PC5372-100G PC5372-25G PC5372-5G
APOSPC5372-100GEA 412 EUR
APOSPC5372-100G APOSPC5372-25G APOSPC5372-5G
1H,1H,7H-Dodecafluoroheptyl-p-toluenesulphonate 95%
1H,1H,7H-Dodecafluoroheptyl-p-toluenesulphonate
Formula: C₁₄H₁₀F₁₂O₃S
MW: 486.28 g/mol
Boiling Pt: 126 °C/0.5mmHg
Melting Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00490197
CAS Number: 424-16-8