4-Bromo-2-(trifluoromethyl)benzenesulphonamide
Supplier: Apollo Scientific
Warning
PC5401-1G PC5401-5G
APOSPC5401-1GEA 23.2 EUR
APOSPC5401-1G APOSPC5401-5G
4-Bromo-2-(trifluoromethyl)benzenesulphonamide
4-Bromo-2-(trifluoromethyl)benzenesulphonamide
Formula: C₇H₅BrF₃NO₂S
Melting Pt: 157…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03094284
CAS Number: 351003-62-8