2-Chloro-5-(trifluoromethyl)pyrimidine 97%
Supplier: Apollo Scientific
Danger
PC5409-1G PC5409-250MG PC5409-5G
APOSPC5409-1GEA 78.3 EUR
APOSPC5409-1G APOSPC5409-250MG APOSPC5409-5G
2-Chloro-5-(trifluoromethyl)pyrimidine 97%
2-Chloro-5-(trifluoromethyl)pyrimidine
Formula: C₅H₂ClF₃N₂
Melting Pt: 47…52 °C
Flash Pt: 71 °C
Storage Temperature: Freezer
MDL Number: MFCD09753482
CAS Number: 69034-12-4
UN: 2811
ADR: 6.1,III