2',3',4',5',6'-Pentafluoroacetophenone 97%
Supplier: Apollo Scientific
Warning
PC5410-25G PC5410-5G PC5410-100G
APOSPC5410-25GEA 107 EUR
APOSPC5410-25G APOSPC5410-5G APOSPC5410-100G
2',3',4',5',6'-Pentafluoroacetophenone 97%
2',3',4',5',6'-Pentafluoroacetophenone
Formula: C₈H₃F₅O
Boiling Pt: 130…133 °C
Density: 1.476 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000296
CAS Number: 652-29-9
EINECS: 211-487-6