4-Carbethoxyhexafluorobutyryl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl hexafluoroglutaryl chloride
PC5439-1G PC5439-25G PC5439-5G
APOSPC5439-1GEA 84.4 EUR
APOSPC5439-1G APOSPC5439-25G APOSPC5439-5G
4-Carbethoxyhexafluorobutyryl chloride 97%
4-Carbethoxyhexafluorobutyryl chloride
Derivatising agent for GC; Superior reagent in quantitation of Amphetamine & Methamphetamine in urine by GC/MS.
Formula: C₇H₅ClF₆O₃
MW: 286.554 g/mol
Boiling Pt: 158…160 °C
Density: 1.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054671
CAS Number: 18381-53-8
UN: 3265
ADR: 8,III