6-Fluoro-m-cresol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-5-methylphenol, 1-Fluoro-2-hydroxy-4-methylbenzene
PC5456-100G PC5456-25G PC5456-5G
APOSPC5456-100GEA 220 EUR
APOSPC5456-100G APOSPC5456-25G APOSPC5456-5G
6-Fluoro-m-cresol 98%
6-Fluoro-m-cresol
Formula: C₇H₇FO
Boiling Pt: 172…174 °C
Density: 1.155 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00190101
CAS Number: 63762-79-8