1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,2,2-Tetrafluoroethyl2,2,3,3-tetrafluoropropylether
PC5459-100G PC5459-25G
APOSPC5459-100GEA 72.5 EUR
APOSPC5459-100G APOSPC5459-25G
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether 99%
1,1,2,2-Tetrafluoroethyl-2,2,3,3-tetrafluoropropyl ether
Formula: C₅H₄F₈O
MW: 232.07 g/mol
Boiling Pt: 93.2 °C
Density: 1.5323 g/cm³ (25 °C)
Flash Pt: 27.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00155961
CAS Number: 16627-68-2
UN: 3271
ADR: 3 III