2-Amino-4-fluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC5484-1G PC5484-25G PC5484-5G
APOSPC5484-1GEA 21.9 EUR
APOSPC5484-1G APOSPC5484-25G APOSPC5484-5G
2-Amino-4-fluorobenzonitrile 98%
2-Amino-4-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.1300 g/mol
Melting Pt: 110…111 °C
Storage Temperature: Ambient
MDL Number: MFCD09955451
CAS Number: 80517-22-2
UN: 3439
ADR: 6.1 III