1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,6-Trifluoro-1-bromo-3,5-dichlorobenzene
PC5512-5G PC5512-1G
APOSPC5512-5GEA 0 EUR
APOSPC5512-5G APOSPC5512-1G
1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene
1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene
Formula: C₆BrCl₂F₃
MW: 279.88 g/mol
Melting Pt: 58…59.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03424469
CAS Number: 24812-13-3