Methyl-2-(trifluoromethyl)acrylate
Supplier: Apollo Scientific
Danger
PC5517-1G PC5517-5G
APOSPC5517-1GEA 90.5 EUR
APOSPC5517-1G APOSPC5517-5G
Methyl-2-(trifluoromethyl)acrylate
Methyl-2-(trifluoromethyl)acrylate
Formula: C₅H₅F₃O₂
Boiling Pt: 104…105 °C
Density: 1.262 g/cm³ (25 °C)
Flash Pt: 22.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068659
CAS Number: 382-90-1
UN: 1993
ADR: 3,II