Methyl-3,3,3-trifluoropropanate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3,3,3-Trifluoropropionate, 3,3,3-Trifluoropropionic acid methyll ester
PC5518-1G PC5518-5G
APOSPC5518-1GEA 21.9 EUR
APOSPC5518-1G APOSPC5518-5G
Methyl-3,3,3-trifluoropropanate
Methyl-3,3,3-trifluoropropanate
Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD02262206
CAS Number: 18830-44-9
UN: 3272
ADR: 3 III