2,4,6-Trifluorophenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: (2,4,6-Trifluorophenyl)metaboric acid, 2,4,6-Trifluorobenzeneboronic acid, Dihydroxy(2,4,6-trifluorophenyl)borane
PC5536-1G PC5536-5G PC5536-25G
APOSPC5536-1GEA 21.9 EUR
APOSPC5536-1G APOSPC5536-5G APOSPC5536-25G
2,4,6-Trifluorophenylboronic acid
2,4,6-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
Melting Pt: 228…235 °C
Storage Temperature: Ambient
MDL Number: MFCD01863169
CAS Number: 182482-25-3