2,6-Difluoro-4-methoxyaniline
Supplier: Apollo Scientific
Danger
Synonyms: 4-Amino-3,5-difluoroanisole
PC5543-1G PC5543-5G PC5543-10G
APOSPC5543-1GEA 40.6 EUR
APOSPC5543-1G APOSPC5543-5G APOSPC5543-10G
2,6-Difluoro-4-methoxyaniline
2,6-Difluoro-4-methoxyaniline
Formula: C₇H₇F₂NO
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD04115922
CAS Number: 151414-47-0
UN: 2811
ADR: 6.1,III