1-Iodo-1H,1H-perfluorohexane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Iodo-1H,1H-undecafluorohexane
PC5587-1G PC5587-5G
APOSPC5587-1GEA 178 EUR
APOSPC5587-1G APOSPC5587-5G
1-Iodo-1H,1H-perfluorohexane 97%
1-Iodo-1H,1H-perfluorohexane
Formula: C₆H₂F₁₁I
MW: 409.97 g/mol
Boiling Pt: 133 °C
Density: 2.018 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04038314
CAS Number: 335-50-2