3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Warning
PC5588-1G PC5588-5G PC5588-25G
APOSPC5588-1GEA 21.9 EUR
APOSPC5588-1G APOSPC5588-5G APOSPC5588-25G
3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%
3-Amino-2-chloro-6-(trifluoromethyl)pyridine
Formula: C6H4ClF3N2
MW: 196.5600 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01862661
CAS Number: 117519-09-2
UN: 2811
ADR: 6.1 III