1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC56077-1G PC56077-250MG
APOSPC56077-1GEA 133 EUR
APOSPC56077-1G APOSPC56077-250MG
1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
1-(1,1-Difluoroethyl)-2-fluorobenzene
Formula: C₈H₇F₃
MW: 160.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845814
CAS Number: 1138445-14-3