4-[(Trifluoromethyl)sulphonyl]aniline 97%
Supplier: Apollo Scientific
Warning
PC5612-1G PC5612-500MG PC5612-5G
APOSPC5612-1GEA 46.4 EUR
APOSPC5612-1G APOSPC5612-500MG APOSPC5612-5G
4-[(Trifluoromethyl)sulphonyl]aniline 97%
4-[(Trifluoromethyl)sulphonyl]aniline
Formula: C₇H₆F₃NO₂S
Melting Pt: 96…97 °C
Storage Temperature: Ambient
CAS Number: 473-27-8