2-Chloro-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC5613-25G PC5613-5G PC5613-100G
APOSPC5613-25GEA 49.3 EUR
APOSPC5613-25G APOSPC5613-5G APOSPC5613-100G
2-Chloro-5-(trifluoromethyl)benzaldehyde 97%
2-Chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208.57
Boiling Pt: 42…44 °C/1.5mmHg
Density: 1.435 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00134430
CAS Number: 82386-89-8