Methyl-4-(trifluoromethoxy)benzoate 97%
Supplier: Apollo Scientific
Warning
PC5623-1G PC5623-25G PC5623-5G
APOSPC5623-1GEA 21.9 EUR
APOSPC5623-1G APOSPC5623-25G APOSPC5623-5G
Methyl-4-(trifluoromethoxy)benzoate 97%
Methyl 4-(trifluoromethoxy)benzoate
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Boiling Pt: 88…90 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00041006
CAS Number: 780-31-4