2-Amino-4-(2,4-difluorophenyl)thiazole
Supplier: Apollo Scientific
Warning
PC5624-1G PC5624-250MG
APOSPC5624-1GEA 52.2 EUR
APOSPC5624-1G APOSPC5624-250MG
2-Amino-4-(2,4-difluorophenyl)thiazole
2-Amino-4-(2,4-difluorophenyl)thiazole
Formula: C₉H₆F₂N₂S
MW: 212.22 g/mol
Melting Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00472891
CAS Number: 105512-80-9