2,4-Dichloro-6-fluorobenzoic acid 95+%
Supplier: Apollo Scientific
PC56240-1G PC56240-5G
APOSPC56240-1GEA 106 EUR
APOSPC56240-1G APOSPC56240-5G
2,4-Dichloro-6-fluorobenzoic acid 95+%
2,4-Dichloro-6-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
MW: 209.0019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12404936
CAS Number: 904285-09-2