4,4,5,5,5-Pentafluoropentan-1-ol 96%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,5,5,5-Pentafluoropentanol
PC5626-25G PC5626-5G PC5626-1G
APOSPC5626-25GEA 94.4 EUR
APOSPC5626-25G APOSPC5626-5G APOSPC5626-1G
4,4,5,5,5-Pentafluoropentan-1-ol 96%
4,4,5,5,5-Pentafluoropentan-1-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 134…135 °C
Melting Pt: –78 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00153224
CAS Number: 148043-73-6
EINECS: 421-360-9