3-Fluoro-4-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
PC5631-10G PC5631-1G PC5631-5G
APOSPC5631-10GEA 698 EUR
APOSPC5631-10G APOSPC5631-1G APOSPC5631-5G
3-Fluoro-4-(trifluoromethyl)pyridine 98%
3-Fluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
Boiling Pt: 37 °C/10mmHg
Density: 1.391 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD03789250
CAS Number: 113770-87-9