2-(Trifluoromethyl)-4-quinolinol 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline
PC5632-1G PC5632-5G
APOSPC5632-1GEA 21.9 EUR
APOSPC5632-1G APOSPC5632-5G
2-(Trifluoromethyl)-4-quinolinol 95+%
2-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MW: 213.1560 g/mol
Melting Pt: 198…203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00153196
CAS Number: 1701-18-4