1-[Bis(4-fluorophenyl)methyl]piperazine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(4,4'-Difluorodiphenylmethyl)piperazine, 1-(4,4'-Difluorobenzhydryl)piperazine, 4,4-Difluorobenzhydrylpiperazine
PC5636-100G PC5636-25G PC5636-5G
APOSPC5636-100GEA 145 EUR
APOSPC5636-100G APOSPC5636-25G APOSPC5636-5G
1-[Bis(4-fluorophenyl)methyl]piperazine 98%
1-[Bis(4-fluorophenyl)methyl]piperazine
Formula: C₁₇H₁₈F₂N₂
MW: 288.34 g/mol
Boiling Pt: 130 °C
Melting Pt: 88…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00038660
CAS Number: 27469-60-9
EINECS: 248-476-0
UN: 2811
ADR: 6.1,III