(Pentafluorophenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,4,5,6-Pentafluorophenylacetic acid
PC5650-25G PC5650-5G PC5650-1G
APOSPC5650-25GEA 545 EUR
APOSPC5650-25G APOSPC5650-5G APOSPC5650-1G
(Pentafluorophenyl)acetic acid 98%
(Pentafluorophenyl)acetic acid
Formula: C₈H₃F₅O₂
MW: 226.1 g/mol
Melting Pt: 107…110 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004316
CAS Number: 653-21-4
EINECS: 211-497-0