4-Chloro-2,6-difluorophenol 97%
Supplier: Apollo Scientific
Danger
PC5657-1G PC5657-25G PC5657-5G
APOSPC5657-1GEA 21.9 EUR
APOSPC5657-1G APOSPC5657-25G APOSPC5657-5G
4-Chloro-2,6-difluorophenol 97%
4-Chloro-2,6-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD06660242
CAS Number: 164790-68-5
UN: 3261
ADR: 8 III