4'-Fluoro-3'-nitroacetophenone
Supplier: Apollo Scientific
Warning
PC5672-25G PC5672-5G
APOSPC5672-25GEA 48 EUR
APOSPC5672-25G APOSPC5672-5G
4'-Fluoro-3'-nitroacetophenone
4'-Fluoro-3'-nitroacetophenone
Formula: C₈H₆FNO₃
MW: 183.14 g/mol
Boiling Pt: 118…120 °C/0.8mmHg
Melting Pt: 47…51 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00115369
CAS Number: 400-93-1