4-Bromo-2-fluorotoluene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-fluoro-4-methylbenzene
PC56731-100G PC56731-1KG PC56731-500G
APOSPC56731-100GEA 26.1 EUR
APOSPC56731-100G APOSPC56731-1KG APOSPC56731-500G
4-Bromo-2-fluorotoluene 95%
4-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8