4-Bromo-3-chloro-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC56867-1G PC56867-5G PC56867-25G
APOSPC56867-1GEA 132 EUR
APOSPC56867-1G APOSPC56867-5G APOSPC56867-25G
4-Bromo-3-chloro-2-fluorobenzoic acid 98%
4-Bromo-3-chloro-2-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16622238
CAS Number: 194804-94-9