2-Bromo-1-chloro-3-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
PC56881-100G PC56881-25G PC56881-5G
APOSPC56881-100GEA 255 EUR
APOSPC56881-100G APOSPC56881-25G APOSPC56881-5G
2-Bromo-1-chloro-3-fluorobenzene 98%
2-Bromo-1-chloro-3-fluorobenzene
Formula: C₆H₃BrClF
MW: 209.4434 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878161
CAS Number: 309721-44-6