2-(3-Fluorophenyl)ethanol 98%
Supplier: Apollo Scientific
Warning
PC5701-1G PC5701-25G PC5701-5G PC5701-100G
APOSPC5701-1GEA 14.7 EUR
APOSPC5701-1G APOSPC5701-25G APOSPC5701-5G APOSPC5701-100G
2-(3-Fluorophenyl)ethanol 98%
2-(3-Fluorophenyl)ethanol
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 90 °C/3mmHg
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00045998
CAS Number: 52059-53-7