1,1,1,2,2-Pentafluoropropane 98%
Supplier: Apollo Scientific
Warning
PC5702-25G PC5702-5G
APOSPC5702-25GEA 951 EUR
APOSPC5702-25G APOSPC5702-5G
1,1,1,2,2-Pentafluoropropane 98%
1,1,1,2,2-Pentafluoropropane
Formula: C₃H₃F₅
Boiling Pt: –18 °C
Melting Pt: –81 °C
Density: 1.176 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069108
CAS Number: 1814-88-6
UN: 3163
ADR: 2.2,