2,6-Difluoro-3-methoxyaniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-2,4-difluoroanisole
PC57026-5G PC57026-1G
APOSPC57026-5GEA 799 EUR
APOSPC57026-5G APOSPC57026-1G
2,6-Difluoro-3-methoxyaniline 98%
2,6-Difluoro-3-methoxyaniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115898
CAS Number: 144851-62-7
UN: 2810
ADR: 6.1 III