Methyl-3-fluorobenzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluorobenzoic acid emthyl ester, Methyl 3-fluorobenzoate
PC5734-100G PC5734-25G
APOSPC5734-100GEA 49.3 EUR
APOSPC5734-100G APOSPC5734-25G
Methyl-3-fluorobenzoate 97%
Methyl 3-fluorobenzoate
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 197 °C
Melting Pt: –10 °C
Density: 1.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094302
CAS Number: 455-68-5