Pentafluoropropionamide 97%
Supplier: Apollo Scientific
|
Warning
|
PC5735-100G
PC5735-25G
APOSPC5735-100GEA
944
EUR
APOSPC5735-100G
APOSPC5735-25G
Pentafluoropropionamide 97%
Pentafluoropropionamide
Formula:
C₃H₂F₅NO MW: 163.05 g/mol Melting Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039771 CAS Number: 354-76-7 EINECS: 206-569-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...