2-Fluoro-3-pyridineboronic acid pinacol ester 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoropyridine-3-boronic acid pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PC5755-1G PC5755-25G PC5755-5G
APOSPC5755-1GEA 21.9 EUR
APOSPC5755-1G APOSPC5755-25G APOSPC5755-5G
2-Fluoro-3-pyridineboronic acid pinacol ester 98%
2-Fluoro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD06798245
CAS Number: 452972-14-4